1,4-Dioxa-7-thiaspiro[4.4]nonane ≥97%

Supplier: Matrix Scientific

114962-1G
102895-100EA 570.77 USD
102895-100
1,4-Dioxa-7-thiaspiro[4.4]nonane ≥97%
1,4-Dioxa-7-thiaspiro[4.4]nonane

Hazard: Irritant

Formula: C₆H₁₀O₂S
MDL Number: MFCD00068021
CAS Number: 176-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR