Dimethyl-3-cyclopentene-1,1-dicarboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Dimethyl cyclopent-3-ene-1,1-dicarboxylate
114720-10G
114720-1G
102894-594EA
253.85
USD
102894-594
102894-596
Dimethyl-3-cyclopentene-1,1-dicarboxylate ≥97%
Dimethyl-3-cyclopentene-1,1-dicarboxylate
Hazard: Irritant
Formula:
C₉H₁₂O₄ MW: 184.19 g/mol |
MDL Number:
MFCD00154460 CAS Number: 84646-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...