Methyl-5-(benzyloxy)-1H-indole-2-carboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 5-(benzyloxy)-1H-indole-2-carboxylate

114698-1G
102894-550EA 641.54 USD
102894-550
Methyl-5-(benzyloxy)-1H-indole-2-carboxylate ≥97%
Methyl-5-(benzyloxy)-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₇H₁₅NO₃
MW: 281.31 g/mol
MDL Number: MFCD07364535
CAS Number: 55581-41-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR