Methyl-2-chloro-5-(trifluoromethyl)nicotinate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 2-chloro-5-(trifluoromethyl)nicotinate

114662-1G
102894-456EA 1298.46 USD
102894-456
Methyl-2-chloro-5-(trifluoromethyl)nicotinate ≥97%
Methyl-2-chloro-5-(trifluoromethyl)nicotinate

Hazard: Irritant

Formula: C₈H₅ClF₃NO₂
MW: 239.58 g/mol
MDL Number: MFCD22559950
CAS Number: 1360934-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR