Methyl-3-nitrophenylacetate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-Nitrophenylacetate

114647-1G 114647-10G
102894-424EA 350.77 USD
102894-424 102894-426
Methyl-3-nitrophenylacetate ≥97%
Methyl-3-nitrophenylacetate

Hazard: Irritant

Formula: C₉H₉NO₄
MW: 195.18 g/mol
MDL Number: MFCD08669939
CAS Number: 10268-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR