Methyl-4-bromo-2,6-dimethylbenzoate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl 4-bromo-2,6-dimethylbenzoate
114624-1G
102898-444EA
884.62
USD
102898-444
Methyl-4-bromo-2,6-dimethylbenzoate ≥97%
Methyl-4-bromo-2,6-dimethylbenzoate
Hazard: Irritant
Formula:
C₁₀H₁₁BrO₂ MW: 243.11 g/mol |
MDL Number:
MFCD11227152 CAS Number: 90841-46-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...