1H-Indazole-3-carbonyl chloride ≥97%

Supplier: Matrix Scientific

114602-1G
102894-376EA 958.46 USD
102894-376
1H-Indazole-3-carbonyl chloride ≥97%
1H-Indazole-3-carbonyl chloride

Hazard: Irritant

Formula: C₈H₅ClN₂O
MDL Number: MFCD11846517
CAS Number: 72083-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR