4-Amino-2-methylpyrimidine-5-carboxylic acid ≥97%

Supplier: Matrix Scientific

114538-1G
102894-250EA 607.69 USD
102894-250
4-Amino-2-methylpyrimidine-5-carboxylic acid ≥97%
4-Amino-2-methylpyrimidine-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₇N₃O₂
MDL Number: MFCD00186151
CAS Number: 769-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR