[(4-Amino-3,5-dichloro-6-fluoropyridin-2-yl)oxy]acetic acid ≥97%

Supplier: Matrix Scientific

114523-1G
102894-220EA 253.85 USD
102894-220
[(4-Amino-3,5-dichloro-6-fluoropyridin-2-yl)oxy]acetic acid ≥97%
[(4-Amino-3,5-dichloro-6-fluoropyridin-2-yl)oxy]acetic acid

Hazard: Irritant

Formula: C₇H₅Cl₂FN₂O₃
MDL Number: MFCD01311822
CAS Number: 69377-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR