2,4-Dichloronicotinoyl chloride ≥97%

Supplier: Matrix Scientific

114482-1G
102894-136EA 350.77 USD
102894-136
2,4-Dichloronicotinoyl chloride ≥97%
2,4-Dichloronicotinoyl chloride

Hazard: Irritant

Formula: C₆H₂Cl₃NO
MDL Number: MFCD19686986
CAS Number: 607351-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR