2-Phenoxybenzoyl chloride ≥97%

Supplier: Matrix Scientific

114472-1G
102894-114EA 738.46 USD
102894-114
2-Phenoxybenzoyl chloride ≥97%
2-Phenoxybenzoyl chloride

Hazard: Corrosive/Stored under Argon/Moisture Sensitive

Formula: C₁₃H₉ClO₂
Melting Pt: 37…39 °C
MDL Number: MFCD03424711
CAS Number: 40501-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR