N-Methyl-1-(5-methyl-2-thienyl)methanamine ≥97%

Supplier: Matrix Scientific

114416-1G
102893-990EA 400 USD
102893-990
N-Methyl-1-(5-methyl-2-thienyl)methanamine ≥97%
N-Methyl-1-(5-methyl-2-thienyl)methanamine

Hazard: Irritant

Formula: C₇H₁₁NS
MDL Number: MFCD04577955
CAS Number: 82230-49-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR