3-(1H-Pyrrol-1-yl)benzylamine ≥97%

Supplier: Matrix Scientific

114410-1G
102893-978EA 813.85 USD
102893-978
3-(1H-Pyrrol-1-yl)benzylamine ≥97%
3-(1H-Pyrrol-1-yl)benzylamine

Hazard: Irritant

Formula: C₁₁H₁₂N₂
MDL Number: MFCD03086143
CAS Number: 368869-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR