4-(4-Methylpiperazin-1-yl)benzoyl chloride ≥97%

Supplier: Matrix Scientific

114309-1G
102898-400EA 958.46 USD
102898-400
4-(4-Methylpiperazin-1-yl)benzoyl chloride ≥97%
4-(4-Methylpiperazin-1-yl)benzoyl chloride

Hazard: Irritant

Formula: C₁₂H₁₅ClN₂O
MDL Number: MFCD20483877
CAS Number: 401495-67-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR