4-Chloro-2-methylbenzoyl chloride ≥97%

Supplier: Matrix Scientific

114306-1G
102898-394EA 641.54 USD
102898-394
4-Chloro-2-methylbenzoyl chloride ≥97%
4-Chloro-2-methylbenzoyl chloride

Hazard: Irritant

Formula: C₈H₆Cl₂O
MDL Number: MFCD11111061
CAS Number: 21900-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR