4-Bromo-2,6-dimethylbenzoyl chloride ≥97%

Supplier: Matrix Scientific

114301-1G
102893-770EA 958.46 USD
102893-770
4-Bromo-2,6-dimethylbenzoyl chloride ≥97%
4-Bromo-2,6-dimethylbenzoyl chloride

Hazard: Irritant

Formula: C₉H₈BrClO
MDL Number: MFCD27928216
CAS Number: 74346-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR