(6S)-5,6-Dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-4-one-7,7-dioxide ≥97%

Supplier: Matrix Scientific

114255-1G
102893-662EA 958.46 USD
102893-662
(6S)-5,6-Dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-4-one-7,7-dioxide ≥97%
(6S)-5,6-Dihydro-6-methyl-4H-thieno[2,3-b]thiopyran-4-one-7,7-dioxide

Hazard: Irritant

Formula: C₈H₈O₃S₂
MDL Number: MFCD13185942
CAS Number: 148719-91-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR