1H-Pyrrolo[2,3-b]pyridine-3-carbohydrazide ≥97%

Supplier: Matrix Scientific

114251-1G
102893-654EA 813.85 USD
102893-654
1H-Pyrrolo[2,3-b]pyridine-3-carbohydrazide ≥97%
1H-Pyrrolo[2,3-b]pyridine-3-carbohydrazide

Hazard: Irritant

Formula: C₈H₈N₄O
MDL Number: MFCD19690324
CAS Number: 310887-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR