4-Chloropyridine-3-sulfonic acid ≥97%

Supplier: Matrix Scientific

114220-1G
102893-588EA 641.54 USD
102893-588
4-Chloropyridine-3-sulfonic acid ≥97%
4-Chloropyridine-3-sulfonic acid

Hazard: Irritant

Formula: C₅H₄ClNO₃S
MDL Number: MFCD01928012
CAS Number: 51498-38-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR