2-(4-Bromo-3-chlorophenyl)acetonitrile ≥97%

Supplier: Matrix Scientific

114191-1G
102893-526EA 813.85 USD
102893-526
2-(4-Bromo-3-chlorophenyl)acetonitrile ≥97%
2-(4-Bromo-3-chlorophenyl)acetonitrile

Hazard: Irritant

Formula: C₈H₅BrClN
MDL Number: MFCD18390590
CAS Number: 1259023-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR