5-Hydroxy-2,1,3-benzothiadiazole ≥97%

Supplier: Matrix Scientific

Synonyms: Benzo[c][1,2,5]thiadiazol-5-ol

114118-1G
102893-348EA 813.85 USD
102893-348
5-Hydroxy-2,1,3-benzothiadiazole ≥97%
5-Hydroxy-2,1,3-benzothiadiazole

Hazard: Irritant

Formula: C₆H₄N₂OS
MW: 152.18 g/mol
MDL Number: MFCD00457955
CAS Number: 768-10-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR