2,4-Dichloro-6-phenylpyrimidine ≥97%

Supplier: Matrix Scientific

114075-1G
102893-258EA 544.62 USD
102893-258
2,4-Dichloro-6-phenylpyrimidine ≥97%
2,4-Dichloro-6-phenylpyrimidine

Hazard: Irritant

Formula: C₁₀H₆Cl₂N₂
MDL Number: MFCD00234591
CAS Number: 26032-72-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR