1-(4-Iodophenyl)hexan-1-one ≥97%

Supplier: Matrix Scientific

114040-1G
102893-192EA 641.54 USD
102893-192
1-(4-Iodophenyl)hexan-1-one ≥97%
1-(4-Iodophenyl)hexan-1-one

Hazard: Irritant

Formula: C₁₂H₁₅IO
MDL Number: MFCD11210528
CAS Number: 160565-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR