Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate ≥98%

Supplier: Matrix Scientific

Synonyms: Ethyl 3-cyano-2-hydroxy-6-methylisonicotinate

113970-1G
102898-334EA 170.77 USD
102898-334
Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate ≥98%
Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate

Hazard: Irritant

Formula: C₁₀H₁₀N₂O₃
MW: 206.2 g/mol
MDL Number: MFCD00052274
CAS Number: 18619-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR