Ethyl (6-aminopyridin-2-yl)acetate ≥97%

Supplier: Matrix Scientific

113965-1G
102898-102EA 620 USD
102898-102
Ethyl (6-aminopyridin-2-yl)acetate ≥97%
Ethyl (6-aminopyridin-2-yl)acetate

Hazard: Irritant

Formula: C₉H₁₂N₂O₂
MDL Number: MFCD09757476
CAS Number: 71469-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR