Ethyl-4-amino-3,5-dichlorobenzoate ≥97%

Supplier: Matrix Scientific

113935-1G
102898-256EA 121.54 USD
102898-256
Ethyl-4-amino-3,5-dichlorobenzoate ≥97%
Ethyl 4-amino-3,5-dichlorobenzoate

Hazard: Irritant

Formula: C₉H₉Cl₂NO₂
MDL Number: MFCD00016636
CAS Number: 74878-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR