Methyl-3-methoxy-5-methylbenzoate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl 3-methoxy-5-methylbenzoate

113923-1G
102893-004EA 884.62 USD
102893-004
Methyl-3-methoxy-5-methylbenzoate ≥97%
Methyl 3-methoxy-5-methylbenzoate

Hazard: Irritant

Formula: C₁₀H₁₂O₃
MW: 180.2000 g/mol
MDL Number: MFCD09833214
CAS Number: 108593-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR