2-Methyl-1,3-oxazole-4-carbonyl chloride ≥97%

Supplier: Matrix Scientific

113915-1G
102892-988EA 958.46 USD
102892-988
2-Methyl-1,3-oxazole-4-carbonyl chloride ≥97%
2-Methyl-1,3-oxazole-4-carbonyl chloride

Hazard: Corrosive

Formula: C₅H₄ClNO₂
MDL Number: MFCD13173537
CAS Number: 62348-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR