4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97%

Supplier: Matrix Scientific

Synonyms: 18-Nitro-2,3,5,6,8,9,11,12,14,15-decahydrobenzo[b][1,4,7,10,13,16]hexaoxacyclooctadecine

113738-1G
102892-718EA 253.85 USD
102892-718
4'-Nitrobenzo-18-crown-6 (4'-nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane) ≥97%
4'-Nitrobenzo-18-crown-6 (4'-Nitrobenzo-1,4,7,10,13,16-hexoxacyclooctadecane)

Hazard: Irritant

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR