Cyclooctane-1,5-dione ≥97%

Supplier: Matrix Scientific

113700-1G
102892-648EA 2303.08 USD
102892-648
Cyclooctane-1,5-dione ≥97%
Cyclooctane-1,5-dione

Hazard: Irritant

Formula: C₈H₁₂O₂
MDL Number: MFCD00101719
CAS Number: 1489-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR