(4-Methyl-6,7-dihydro-4H-thieno-[3,2-c]pyran-4-yl)acetic acid ≥97%

Supplier: Matrix Scientific

113647-1G
102897-970EA 738.46 USD
102897-970
(4-Methyl-6,7-dihydro-4H-thieno-[3,2-c]pyran-4-yl)acetic acid ≥97%
(4-Methyl-6,7-dihydro-4H-thieno-[3,2-c]pyran-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₂O₃S
MW: 212.27 g/mol
MDL Number: MFCD01850960
CAS Number: 57153-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR