3,5-Difluorosalicylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 3,5-Difluoro-2-hydroxybenzoic acid

113631-1G
102897-944EA 400 USD
102897-944
3,5-Difluorosalicylic acid ≥97%
3,5-Difluorosalicylic acid

Hazard: Irritant

Formula: C₇H₄F₂O₃
MW: 174.10
MDL Number: MFCD06203609
CAS Number: 84376-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR