1-Adamantylacetylene

Supplier: Matrix Scientific

113472-1G
102892-216EA 1129.23 USD
102892-216
1-Adamantylacetylene
1-Adamantylacetylene

Hazard: Irritant

Formula: C₁₂H₁₆
MDL Number: MFCD08061913
CAS Number: 40430-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR