(4-Chlorothiazol-2-yl)acetic acid

Supplier: Matrix Scientific

113463-1G
102897-848EA 1978.46 USD
102897-848
(4-Chlorothiazol-2-yl)acetic acid
(4-Chlorothiazol-2-yl)acetic acid

Hazard: Irritant

Formula: C₅H₄ClNO₂S
MDL Number: MFCD23106222
CAS Number: 29676-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR