6-Boc-2-hydroxy-6-azaspiro[3.5]nonane

Supplier: Matrix Scientific

113441-1G
102892-162EA 1600 USD
102892-162
6-Boc-2-hydroxy-6-azaspiro[3.5]nonane
6-Boc-2-hydroxy-6-azaspiro[3.5]nonane

Hazard: Irritant

Formula: C₁₃H₂₃NO₃
MDL Number: MFCD23106479
CAS Number: 1419101-54-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR