2-Amino-4-ethynylpyrimidine

Supplier: Matrix Scientific

113381-1G
102892-014EA 2181.54 USD
102892-014
2-Amino-4-ethynylpyrimidine
2-Amino-4-ethynylpyrimidine

Hazard: Irritant

Formula: C₆H₅N₃
MDL Number: MFCD12755763
CAS Number: 1207175-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR