1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile

Supplier: Matrix Scientific

113321-1G
102891-926EA 1309.23 USD
102891-926
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile

Hazard: Irritant

Formula: C₇H₄N₄
MDL Number: MFCD13176785
CAS Number: 956010-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR