5,8-Dioxaspiro[3.4]octane-2-methanol

Supplier: Matrix Scientific

113313-1G 113313-5G
102897-734EA 436.92 USD
102897-734 102897-736
5,8-Dioxaspiro[3.4]octane-2-methanol
5,8-Dioxaspiro[3.4]octane-2-methanol

Hazard: Irritant

Formula: C₇H₁₂O₃
MDL Number: MFCD23106218
CAS Number: 545882-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR