Ethyl-4-azaindole-6-carboxylate
Supplier: Matrix Scientific
113309-1G
102891-918EA
1687.69
USD
102891-918
Ethyl-4-azaindole-6-carboxylate
Ethyl-4-azaindole-6-carboxylate
Hazard: Irritant
Formula:
C₁₀H₁₀N₂O₂ |
MDL Number:
MFCD18416564 CAS Number: 1261885-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...