3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride

Supplier: Matrix Scientific

113282-1G
102891-826EA 1687.69 USD
102891-826
3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride
3-Amino-1-(trifluoromethyl)cyclobutan-1-ol hydrochloride

Hazard: Irritant

Formula: C₅H₉ClF₃NO
MDL Number: MFCD22392108
CAS Number: 1408075-12-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR