Methyl-1-boc-3-ethylazetidine-3-carboxylate

Supplier: Matrix Scientific

113281-1G
102891-824EA 1018.46 USD
102891-824
Methyl-1-boc-3-ethylazetidine-3-carboxylate
Methyl-1-boc-3-ethylazetidine-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₂₁NO₄
MW: 243.31 g/mol
MDL Number: MFCD18374998
CAS Number: 1262410-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR