1-Boc-2-methyl-D-proline
Supplier: Matrix Scientific
Synonyms:
(R)-1-(tert-Butoxycarbonyl)-2-methylpyrrolidine-2-carboxylic acid
113279-1G
102891-820EA
1018.46
USD
102891-820
1-Boc-2-methyl-D-proline
1-Boc-2-methyl-D-proline
Hazard: Irritant
Formula:
C₁₁H₁₉NO₄ MW: 229.28 g/mol |
MDL Number:
MFCD06797554 CAS Number: 166170-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...