(S)-3-(Cbz-amino)piperidine

Supplier: Matrix Scientific

Synonyms: (S)-Benzyl piperidin-3-ylcarbamate

113278-1G
102891-818EA 1018.46 USD
102891-818
(S)-3-(Cbz-amino)piperidine
(S)-3-(Cbz-amino)piperidine

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MW: 234.3 g/mol
MDL Number: MFCD03426377
CAS Number: 478646-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR