Methyl-2-(boc-amino)-2-(oxetan-3-yl)acetate

Supplier: Matrix Scientific

113258-1G
102897-802EA 2589.23 USD
102897-802
Methyl-2-(boc-amino)-2-(oxetan-3-yl)acetate
Methyl-2-(boc-amino)-2-(oxetan-3-yl)acetate

Hazard: Irritant

Formula: C₁₁H₁₉NO₅
MW: 245.28 g/mol
MDL Number: MFCD22628746
CAS Number: 1416323-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR