2-(Boc-amino)-2-(oxetan-3-yl)acetic acid

Supplier: Matrix Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-2-(oxetan-3-yl)acetic acid

113257-1G
102897-800EA 2589.23 USD
102897-800
2-(Boc-amino)-2-(oxetan-3-yl)acetic acid
2-(Boc-amino)-2-(oxetan-3-yl)acetic acid

Hazard: Irritant

Formula: C₁₀H₁₇NO₅
MW: 231.25 g/mol
MDL Number: MFCD21848380
CAS Number: 1408074-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR