Methyl-5-amino-4-methylpyridine-2-carboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 5-amino-4-methylpicolinate

113190-1G
102897-630EA 1309.23 USD
102897-630
Methyl-5-amino-4-methylpyridine-2-carboxylate
Methyl-5-amino-4-methylpyridine-2-carboxylate

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD18803232
CAS Number: 868551-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR