3-Ethynylimidazo[1,2-b]pyridazine

Supplier: Matrix Scientific

113111-1G
102897-708EA 1309.23 USD
102897-708
3-Ethynylimidazo[1,2-b]pyridazine
3-Ethynylimidazo[1,2-b]pyridazine

Hazard: Irritant

Formula: C₈H₅N₃
MDL Number: MFCD17012027
CAS Number: 943320-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR