2-Hydroxymethyl-5-methylpyrimidine

Supplier: Matrix Scientific

113060-1G
102897-660EA 2181.54 USD
102897-660
2-Hydroxymethyl-5-methylpyrimidine
2-Hydroxymethyl-5-methylpyrimidine

Hazard: Irritant

Formula: C₆H₈N₂O
MDL Number: MFCD10697091
CAS Number: 90905-61-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR