3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester

Supplier: Matrix Scientific

113052-1G
102897-594EA 1018.46 USD
102897-594
3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester
3,3-Difluorocyclobutane-1,1-dicarboxylic-1-isopropyl ester

Hazard: Irritant

Formula: C₉H₁₂F₂O₄
MW: 222.19 g/mol
MDL Number: MFCD23105828
CAS Number: 1225532-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR