1-Methyl-3-oxocyclobutane-1-carboxylic acid

Supplier: Matrix Scientific

Synonyms: 1-Methyl-3-oxocyclobutanecarboxylic acid

113202-5G 113202-1G
102891-652EA 2000 USD
102891-652 102891-650
1-Methyl-3-oxocyclobutane-1-carboxylic acid
1-Methyl-3-oxocyclobutane-1-carboxylic acid

Hazard: Irritant

Formula: C₆H₈O₃
MW: 128.13 g/mol
MDL Number: MFCD00604210
CAS Number: 286442-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR